Aluminium isopropoxide ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminiumtriisopropylate, Aluminiumisopropylate, Aluminiumtriisopropoxide, Aluminium isopropoxide

22982-14 22982-22
CAAA22982-14EA 153.1 CAD
CAAA22982-14 CAAA22982-22
Aluminium isopropoxide ≥99.99% (metals basis)
Aluminium isopropoxide

MDL: MFCD00008870


Purity: 99.99+% CAS No.: 555-31-7 Molecular Formula: Al[OCH(CH3)2]3 Formula Weight: 204.25 Physical Form: Powder Density: 1.035 Refractive Index: 1.0346 Sensitivity: Moisture Sensitive Melting Point: 131-135° Boiling Point: 135°/10mm Merck Reference: 14,347 UN No.: 3181 MDL No.: MFCD00008870

Formula: C₉H₂₁AlO₃
MW: 204.25 g/mol
Boiling Pt: 135
Melting Pt: 119
Density: 1.035
Storage Temperature: Ambient
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1 II
Merck Index: 14,347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR