Titanium(IV) chloride ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
22979-14
22979-22
22979-88
CAAA22979-14EA
128.14
CAD
CAAA22979-14
CAAA22979-22
CAAA22979-88
Titanium(IV) chloride ≥99.99% (metals basis)
Titanium (IV) chloride
MDL: MFCD00011267
Fieser: 1,1169 15,317 16,332 17,344 18,359 19,341 20,377 21,425
Purity: 99.99%
CAS No.: 7550-45-0
Molecular Formula: Cl4Ti
Formula Weight: 189.69
Physical Form: Liquid
Density: 1.726
Refractive Index: 1.61
Sensitivity: Moisture Sensitive
Appearance: Colorless to pale yellow
Melting Point: -25°
Boiling Point: 136-137°
Merck Reference: 14,9478
UN No.: 1838
MDL No.: MFCD00011267
Formula:
Cl₄Ti Boiling Pt: 136…137 °C Melting Pt: –25 °C Density: 1.726 Storage Temperature: Ambient |
MDL Number:
MFCD00011267 CAS Number: 7550-45-0 EINECS: 231-441-9 UN: 1838 ADR: 6.1,I Merck Index: 14,9478 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...