Tetraethyl orthosilicate (TEOS) ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetraethoxysilane
22967-18
22967-30
CAAA22967-18EA
135.56
CAD
CAAA22967-18
CAAA22967-30
Tetraethyl orthosilicate (TEOS) ≥99.999% (metals basis)
Tetraethyl orthosilicate (TEOS)
MDL: MFCD00009062
Beilstein Registry No.: 1422225
Purity: 99.999+%
CAS No.: 78-10-4
Molecular Formula: C8H20O4Si
Formula Weight: 208.33
Flash Point: 46°(116°F)
Physical Form: Liquid
Density: 0.934
Refractive Index: 1.3820
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Pleasant
Melting Point: -85°
Boiling Point: 168°
Beilstein Reference: 1422225
Merck Reference: 14,3851
UN No.: 1292
MDL No.: MFCD00009062
Formula:
Si(OC₂H₅)₄ MW: 208.33 g/mol Boiling Pt: 168 °C Melting Pt: –85 °C Density: 0.934 Flash Pt: 46 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009062 CAS Number: 78-10-4 EINECS: 201-083-8 UN: 1292 ADR: 3,III Merck Index: 14,3851 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...