Vanadium(IV) oxide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Vanadium dioxide
22957-09
22957-18
CAAA22957-09EA
144.89
CAD
CAAA22957-09
CAAA22957-18
Vanadium(IV) oxide ≥99% (metals basis)
Vanadium (IV) oxide
MDL: MFCD00049702
Purity: 99%
CAS No.: 12036-21-4
Molecular Formula: O2V, may contain V6O13
Formula Weight: 82.94
Physical Form: -100 Mesh Powder
Density: 4.339
Appearance: Black
Melting Point: 1967°
UN No.: 3285
MDL No.: MFCD00049702
Formula:
O₂V MW: 82.94 Boiling Pt: 2700 Melting Pt: 238 Density: 4.339 Storage Temperature: Ambient |
MDL Number:
MFCD00049702 CAS Number: 12036-21-4 EINECS: 234-841-1 UN: 3285 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...