Dimethyl sulfide 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DMS

22949-AK 22949-K2
CAAA22949-AKEA 62.09 CAD
CAAA22949-AK CAAA22949-K2
Dimethyl sulfide 99+%
Dimethyl sulphide

MDL: MFCD00008562


Purity: 99+% CAS No.: 75-18-3 Molecular Formula: (CH3)2S Formula Weight: 62.14 Flash Point: -48°(-54°F) Physical Form: Liquid Density: 0.845 Refractive Index: 1.4351 Odor: Unpleasant Melting Point: -83° Boiling Point: 38° Merck Reference: 14,6123 UN No.: 1164 MDL No.: MFCD00008562

Formula: C₂H₆S
MW: 62.14 g/mol
Boiling Pt: 36.2
Melting Pt: -98
Density: 0.845
Flash Pt: –48 °C (–54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008562
CAS Number: 75-18-3
EINECS: 200-846-2
UN: 1164
ADR: 3 II
Merck Index: 14,6123

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR