Vanadium(III) acetylacetonate ∼97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
22940-06
22940-14
22940-22
CAAA22940-06EA
41.71
CAD
CAAA22940-06
CAAA22940-14
CAAA22940-22
Vanadium(III) acetylacetonate ∼97%
MDL: MFCD00000033
Purity: 97%
CAS No.: 13476-99-8
Molecular Formula: C15H21O6V
Formula Weight: 348.27
Physical Form: Powder
Sensitivity: Air Sensitive
UN No.: 3285
MDL No.: MFCD00000033
Formula:
C₁₅H₂₁O₆V Boiling Pt: 170 °C (0,05 mmHg) Melting Pt: 185…190 °C Density: ∼1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000033 CAS Number: 13476-99-8 EINECS: 236-759-1 UN: 3285 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...