1-Propanol ≥99% for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
22932-K2
22932-K7
22932-M1
CAAA22932-K2EA
159.58
CAD
CAAA22932-K2
CAAA22932-K7
CAAA22932-M1
1-Propanol ≥99% for HPLC
1-Propanol
MDL: MFCD00002941
Beilstein Registry No.: 1098242
Notes: Filtered through 0.2µ filters. Suitable for spectophotometry, chromatography.
Purity: 99% min
CAS No.: 71-23-8
Molecular Formula: CH3CH2CH2OH
Formula Weight: 60.10
Flash Point: 15°(59°F)
Physical Form: Liquid
Density: 0.804
Refractive Index: 1.3850
Odor: Alcohol-like
Melting Point: -127°
Boiling Point: 97°
Beilstein Reference: 1098242
Merck Reference: 14,7842
UN No.: 1274
MDL No.: MFCD00002941
Formula:
H₃CCH₂CH₂OH Boiling Pt: 97 °C Melting Pt: –127 °C Density: 0.804 Flash Pt: 15 °C (59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002941 CAS Number: 71-23-8 EINECS: 200-746-9 UN: 1274 ADR: 3,II Merck Index: 14,7842 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...