Acetone ≥99.5% for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
22928-M6
22928-K2
22928-K7
22928-M1
CAAA-22928-M6EA
658.86
CAD
CAAA-22928-M6
CAAA22928-K2
CAAA22928-K7
CAAA22928-M1
Acetone ≥99.5% for HPLC
Acetone
MDL: MFCD00008765
Beilstein Registry No.: 635680
Notes: Filtered through 0.2µ filters.
Fieser: 2,13 3,4 6,9
Purity: 99.5+%
CAS No.: 67-64-1
Molecular Formula: C3H6O
Formula Weight: 58.08
Flash Point: -17°(1°F)
Physical Form: Liquid
Density: 0.791
Refractive Index: 1.3590
Appearance: Colorless
Odor: Characteristic
Melting Point: -94°
Boiling Point: 56°
Beilstein Reference: 635680
Merck Reference: 14,66
UN No.: 1090
MDL No.: MFCD00008765
Formula:
CH₃COCH₃ Boiling Pt: 56 °C Melting Pt: –94 °C Density: 0.791 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II REACH: 01-2119471330-49 Merck Index: 14,66 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...