Chloroform ≥99.5% for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
22920-K2
22920-K7
22920-M1
CAAA22920-K2EA
142.99
CAD
CAAA22920-K2
CAAA22920-K7
CAAA22920-M1
Chloroform ≥99.5% for HPLC
Chloroform
MDL: MFCD00000826
Beilstein Registry No.: 1731042
Notes: Filtered through 0.2µ filters.
Fieser: 1,29 3,140 4,22 5,28 8,92 12,517 15,84
Purity: 99.5+% min
CAS No.: 67-66-3
Molecular Formula: CHCl3
Formula Weight: 119.38
Physical Form: Liquid
Density: 1.492
Refractive Index: 1.4460
Odor: Characteristic
Melting Point: -63°
Boiling Point: 60.5-61.5°
Beilstein Reference: 1731042
Merck Reference: 14,2141
UN No.: 1888
MDL No.: MFCD00000826
Formula:
CHCl₃ Boiling Pt: 60.5…61.5 °C Melting Pt: –63 °C Density: 1.492 Storage Temperature: Ambient |
MDL Number:
MFCD00000826 CAS Number: 67-66-3 EINECS: 200-663-8 UN: 1888 ADR: 6.1,III REACH: 05-2114614906-45 Merck Index: 14,2141 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...