Cyclohexane ≥99% for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
22919-K2
22919-K7
22919-M1
CAAA22919-K2EA
130.51
CAD
CAAA22919-K2
CAAA22919-K7
CAAA22919-M1
Cyclohexane ≥99% for HPLC
Cyclohexane
MDL: MFCD00003814
Beilstein Registry No.: 1900225
Notes: Filtered through 0.2µ filters.
Purity: 99% min
CAS No.: 110-82-7
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -18°(-1°F)
Physical Form: Liquid
Density: 0.779
Refractive Index: 1.4260
Appearance: Colorless
Odor: Pungent
Melting Point: 6.5°
Boiling Point: 80-81°
Beilstein Reference: 1900225
Merck Reference: 14,2723
UN No.: 1145
MDL No.: MFCD00003814
Formula:
C₆H₁₂ Boiling Pt: 80…81 °C Melting Pt: 6.5 °C Density: 0.779 Flash Pt: –18 °C (–1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003814 CAS Number: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3,II Merck Index: 14,2723 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...