N,N-Dimethylformamide ≥99.7% for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
22915-K2
22915-K7
22915-M1
22915-M6
22915-Y0
CAAA22915-K2EA
146.94
CAD
CAAA22915-K2
CAAA22915-K7
CAAA22915-M1
CAAA22915-M6
CAAA22915-Y0
N,N-Dimethylformamide ≥99.7% for HPLC
N,N-Dimethylformamide
MDL: MFCD00003284
Beilstein Registry No.: 605365
Notes: Filtered through 0.2µ filters.
Fieser: 1,273 9,182 11,198 12,203 14,148 16,144 18,146 19,137 21,178
Purity: 99.7+%
CAS No.: 68-12-2
Molecular Formula: HCON(CH3)2
Formula Weight: 73.09
Flash Point: 57°(135°F)
Physical Form: Liquid
Density: 0.944
Refractive Index: 1.4310
Odor: Amine-like
pH: 7
Melting Point: -61°
Boiling Point: 153°
Beilstein Reference: 605365
Merck Reference: 14,3243
UN No.: 2265
MDL No.: MFCD00003284
Formula:
HCON(CH₃)₂ Boiling Pt: 153 °C Melting Pt: –61 °C Density: 0.944 Flash Pt: 57 °C (135 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III REACH: 01-2119475605-32 Merck Index: 14,3243 |
|
Specification Test Results
Evaporation residue | 5 ppm |
H2O | 0.0005 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...