1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
22913-K2
22913-K7
22913-M1
CAAA22913-K2EA
183.28
CAD
CAAA22913-K2
CAAA22913-K7
CAAA22913-M1
1,4-Dioxane ≥99% for HPLC
1,4-Dioxane
MDL: MFCD00006571
Beilstein Registry No.: 102551
Notes: Filtered through 0.2µ filters.
Fieser: 1,333
Purity: 99% min
CAS No.: 123-91-1
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: 12°(54°F)
Physical Form: Liquid
Density: 1.034
Refractive Index: 1.4220
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Ether-like
Melting Point: 11.8°
Boiling Point: 100-102°
Beilstein Reference: 102551
Merck Reference: 14,3300
UN No.: 1165
MDL No.: MFCD00006571
Formula:
C₄H₈O₂ Boiling Pt: 100…102 °C Melting Pt: 11.8 °C Density: 1.034 Flash Pt: 12 °C (54 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006571 CAS Number: 123-91-1 EINECS: 204-661-8 UN: 1165 ADR: 3,II Merck Index: 14,3300 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...