Isobutanol 99+% for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
22908-K2
22908-K7
22908-M1
CAAA22908-K2EA
141.09
CAD
CAAA22908-K2
CAAA22908-K7
CAAA22908-M1
Isobutanol 99+% for HPLC
Isobutanol
MDL: MFCD00004740
Beilstein Registry No.: 1730878
Notes: Filtered through 0.2µ filters.
Purity: 99+%
CAS No.: 78-83-1
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 28°(82°F)
Physical Form: Liquid
Density: 0.802
Refractive Index: 1.3960
Appearance: Colorless
Odor: Sweet
pH: 7
Melting Point: -108°
Boiling Point: 107-108°
Beilstein Reference: 1730878
Merck Reference: 14,5131
UN No.: 1212
MDL No.: MFCD00004740
Formula:
(CH₃)₂CHCH₂OH Boiling Pt: 107…108 °C Melting Pt: –108 °C Density: 0.802 Flash Pt: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004740 CAS Number: 78-83-1 EINECS: 201-148-0 UN: 1212 ADR: 3,III Merck Index: 14,5131 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...