o-Xylene ≥96% for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,2-Dimethylbenzene
22902-K2
22902-K7
22902-M1
CAAA22902-K2EA
180.12
CAD
CAAA22902-K2
CAAA22902-K7
CAAA22902-M1
o-Xylene ≥96% for HPLC
o-Xylene
MDL: MFCD00008519
Beilstein Registry No.: 1815558
Notes: Filtered through 0.2µ filters.
Purity: 96% min
CAS No.: 95-47-6
Molecular Formula: C8H10
Formula Weight: 106.17
Flash Point: 32°(89°F)
Physical Form: liquid
Density: 0.877
Refractive Index: 1.5050
Appearance: Colorless
Melting Point: -25°
Boiling Point: 143-145°
Beilstein Reference: 1815558
Merck Reference: 14,10081
UN No.: 1307
MDL No.: MFCD00008519
Formula:
C₈H₁₀ Boiling Pt: 143…145 °C Melting Pt: –25 °C Density: 0.877 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008519 CAS Number: 95-47-6 EINECS: 202-422-2 UN: 1307 ADR: 3,III Merck Index: 14,10081 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...