Potassium molybdate(VI), anhydrous ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
22898-09
22898-18
CAAA22898-09EA
77.42
CAD
CAAA22898-09
CAAA22898-18
Potassium molybdate(VI), anhydrous ≥99.8% (metals basis)
Potassium molybdate(VI)
MDL: MFCD00011370
Purity: 99.8%
CAS No.: 13446-49-6
Molecular Formula: K2MoO4
Formula Weight: 238.14
Physical Form: -200 Mesh Powder
Density: 2.3
Appearance: White
Melting Point: 919°
Merck Reference: 14,7647
MDL No.: MFCD00011370
Formula:
K₂MoO₄ Melting Pt: 919 °C Density: 2.3 Storage Temperature: Ambient |
MDL Number:
MFCD00011370 CAS Number: 13446-49-6 EINECS: 236-599-2 Merck Index: 14,7647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...