Nickel carbonate, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Danger

22897-18 22897-30
CAAA22897-18EA 92.9 CAD
CAAA22897-18 CAAA22897-30
Nickel carbonate, anhydrous 98%
Nickel carbonate

MDL: MFCD00011144


Purity: 98% CAS No.: 3333-67-3 Molecular Formula: CNiO3 Formula Weight: 118.72 Physical Form: -100 Mesh Powder Appearance: Pale purple UN No.: 3077 MDL No.: MFCD00011144

Formula: CNiO₃
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011144
CAS Number: 3333-67-3
EINECS: 222-068-2
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR