Citric acid monohydrate ≥99.5%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

22869-22 22869-36 22869-A1
CAAA22869-22EA 59.25 CAD
CAAA22869-22 CAAA22869-36 CAAA22869-A1
Citric acid monohydrate ≥99.5%
Citric acid monohydrate

MDL: MFCD00149972


Purity: 99.5+% CAS No.: 5949-29-1 Molecular Formula: C6H8O7.H2O Formula Weight: 210.14 (192.13anhy) Physical Form: Granular Density: 1.542 Appearance: White Odor: Odorless Merck Reference: 14,2326 MDL No.: MFCD00149972

Formula: C₆H₈O₇.H₂O
Melting Pt: 135…152 °C
Density: 1.542
Flash Pt: 173.9 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1
Merck Index: 14,2326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR