Crystal violet 90+%, crystals ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
MDL: MFCD00011750
Beilstein Registry No.: 4077708
Purity: 90+%
CAS No.: 548-62-9
Molecular Formula: C25H30ClN3
Formula Weight: 407.99
Physical Form: Powder
Melting Point: ca 215° dec.
Beilstein Reference: 4077708
Merck Reference: 14,4395
UN No.: 3077
MDL No.: MFCD00011750
Formula:
C₂₅H₃₀ClN₃ MW: 407.99 g/mol Melting Pt: ∼ 215 °C (dec.) Density: 1.19 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011750 CAS Number: 548-62-9 EINECS: 208-953-6 UN: 3077 ADR: 9,III Merck Index: 14,4395 |
Specification Test Results
Sensitivity as indicator | Pass test |
Loss on drying | 0.075 |
Absorbance characteristics | Pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...