Cyclohexane 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexahydrobenzene, Hexamethylene
22864-AP
22864-K2
22864-K7
22864-M1
CAAA22864-APEA
69.84
CAD
CAAA22864-AP
CAAA22864-K2
CAAA22864-K7
CAAA22864-M1
Cyclohexane 99+% ACS
Cyclohexane
MDL: MFCD00003814
Beilstein Registry No.: 1900225
Purity: 99+%
CAS No.: 110-82-7
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -18°(-1°F)
Physical Form: Liquid
Density: 0.779
Refractive Index: 1.4260
Odor: Pungent
Melting Point: 6.5°
Boiling Point: 80-81°
Beilstein Reference: 1900225
Merck Reference: 14,2723
UN No.: 1145
MDL No.: MFCD00003814
Formula:
C₆H₁₂ Boiling Pt: 80…81 °C Melting Pt: 6.5 °C Density: 0.779 Flash Pt: –18 °C (–1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003814 CAS Number: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3,II Merck Index: 14,2723 |
|
Specification Test Results
Color (APHA) | 10 |
Substances darkened by sulfuric acid | Pass test |
H2O | 0.02% |
Evaporation residue | 0.002% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...