Silver(II) oxide 98%

Supplier: Thermo Scientific Chemicals
Danger

22695-06 22695-14 22695-22
CAAA22695-06EA 72.68 CAD
CAAA22695-06 CAAA22695-14 CAAA22695-22
Silver(II) oxide 98%
Silver (II) oxide

Manufacturing of silver oxide-zinc alkali batteries


Purity: 98% CAS No.: 1301-96-8 Molecular Formula: AgO Formula Weight: 123.87 Physical Form: Powder Density: 7.44 Appearance: Grey to black Odor: Odorless Melting Point: 100° dec. Solubility: Insoluble in water. Soluble in alkalies, ammonium hydroxide with decomposition and N2 formation Merck Reference: 14,8522 UN No.: 3085 MDL No.: MFCD00044285


Manufacturing of silver oxide-zinc alkali batteries

Formula: AgO
Melting Pt: >100 °C (dec.)
Density: 7.44
Storage Temperature: Ambient
MDL Number: MFCD00044285
CAS Number: 1301-96-8
EINECS: 215-098-2
UN: 3085
ADR: 5.1,II
Merck Index: 14,8522

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR