Dilanthanum trisulfide ≥99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lanthanum (III) sulphide, Lanthanum (III) sulfide

22655-04 22655-09
CAAA22655-04EA 192.76 CAD
CAAA22655-04 CAAA22655-09
Dilanthanum trisulfide ≥99% (REO, rare earth oxide basis)
Dilanthanum trisulphide

MDL: MFCD00016168


Purity: 99% CAS No.: 12031-49-1 Molecular Formula: La2S3 Formula Weight: 374.02 Physical Form: Powder Density: 4.911 Melting Point: 2100° in vac. UN No.: 3134 MDL No.: MFCD00016168

Formula: La₂S₃
MW: 374.01
Melting Pt: 2127
Density: 4.911
Storage Temperature: Ambient
MDL Number: MFCD00016168
CAS Number: 12031-49-1
EINECS: 234-752-8
UN: 3134
ADR: 4.3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR