Indium arsenide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

22651-04 22651-09
CAAA22651-04EA 111.39 CAD
CAAA22651-04 CAAA22651-09
Indium arsenide ≥99% (metals basis)
Indium arsenide

In semiconductor electronics


Purity: 99% CAS No.: 1303-11-3 Molecular Formula: AsIn Formula Weight: 189.74 Physical Form: 5mm & Down Pieces Odor: Odorless Merck Reference: 14,4949 UN No.: 1557 MDL No.: MFCD00016144


In semiconductor electronics

Formula: AsIn
Melting Pt: 936 °C
Density: 5.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016144
CAS Number: 1303-11-3
EINECS: 215-115-3
UN: 1557
ADR: 6.1,III
Merck Index: 14,4949

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR