Calcium aluminate ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals

22646-06 22646-14 22646-22
CAAA22646-06EA 93.69 CAD
CAAA22646-06 CAAA22646-14 CAAA22646-22
Calcium aluminate ≥99% (metals basis)
Calcium aluminate

MDL: MFCD00049722


Purity: 99% CAS No.: 12042-68-1 Molecular Formula: CaAl2O4 Formula Weight: 158.02 Physical Form: -200 Mesh Powder Density: 2.981 Melting Point: 1600° MDL No.: MFCD00049722

Formula: Al₂CaO₄
Melting Pt: 1600 °C
Density: 2,981
Storage Temperature: Ambient
MDL Number: MFCD00049722
CAS Number: 12042-68-1
EINECS: 234-931-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR