Barium aluminate, tech.

Supplier: Thermo Scientific Chemicals
Warning

22642-09 22642-22 22642-36
CAAA22642-09EA 30.02 CAD
CAAA22642-09 CAAA22642-22 CAAA22642-36
Barium aluminate, tech.
Barium aluminate

MDL: MFCD00049716


CAS No.: 12004-04-5 Molecular Formula: BaAl2O4 (approximate formula) Formula Weight: 561.98 Physical Form: Powder Odor: Odorless UN No.: 1564 MDL No.: MFCD00049716

Formula: Al₂BaO₄
Melting Pt: 169.5…170 °C
Density: 4,04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049716
CAS Number: 12004-04-5
EINECS: 234-445-9
UN: 1564
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR