Ethanethiol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
22585-AC
22585-AE
22585-AP
CAAA22585-ACEA
38.55
CAD
CAAA22585-AC
CAAA22585-AE
CAAA22585-AP
Ethanethiol 97%
Ethanethiol
MDL: MFCD00004887
Soluble in alcohol, ether, water
Purity: 97%
CAS No.: 75-08-1
Molecular Formula: C2H6S
Formula Weight: 62.13
Flash Point: -48°(-55°F)
Physical Form: Liquid
Density: 0.84
Refractive Index: 1.4310
Appearance: Colorless
Odor: Unpleasant
Melting Point: -148°
Boiling Point: 35°
Solubility: Soluble in alcohol, ether, water
Beilstein Reference: 773638
Merck Reference: 14,3726
UN No.: 2363
MDL No.: MFCD00004887
Formula:
C₂H₆S Boiling Pt: 35 °C Melting Pt: –148 °C Density: 0.84 Flash Pt: –48 °C (–55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004887 CAS Number: 75-08-1 EINECS: 200-837-3 UN: 2363 ADR: 3,I Merck Index: 14,3726 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...