EDTA tripotassium salt

Supplier: Thermo Scientific Chemicals

Synonyms: Edta-3k, Ethylenediaminetetra-acetic acid tripotassium salt, Tripotassium hydrogen ethylenediaminetetraacetate, Ethylenedinitrilotetra-acetic acid tripotassium salt, Edetate Tripotassium, Edetic acid tripotassium salt, Potassium 2,2'-((2-((carboxylatomethyl)(carboxymethyl)amino)ethyl)azanediyl)diacetate

22425-14 22425-22
CAAA22425-14EA 94.01 CAD
CAAA22425-14 CAAA22425-22
EDTA tripotassium salt
EDTA tripotassium salt

MDL: MFCD00013304


CAS No.: 17572-97-3 Molecular Formula: (KO2CCH2)2NCH2CH2N(CH2CO2H)CH2CO2K Formula Weight: 406.46 Physical Form: Crystalline Odor: Odorless MDL No.: MFCD00013304

Formula: C₁₀H₁₃K₃N₂O₈
MW: 406.52
Storage Temperature: Ambient
MDL Number: MFCD00013304
CAS Number: 17572-97-3
EINECS: 241-543-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR