Chromotropic acid disodium salt dihydrate ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Disodium 4,5-dihydroxynaphthalene-2,7-disulfonate dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate, Disodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate, 4,5-Dihydroxy-2,7-naphthalenedisulfonic acid disodium salt dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, 4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate

22414-06 22414-14
CAAA22414-06EA 59.41 CAD
CAAA22414-06 CAAA22414-14
Chromotropic acid disodium salt dihydrate ACS
Chromotropic acid disodium salt dihydrate

MDL: MFCD00150612 Beilstein Registry No.: 3647189


CAS No.: 5808-22-0 Molecular Formula: C10H6Na2O8S2.2H2O Formula Weight: 400.29 (364.26anhy) Physical Form: Powder Appearance: Off-White to Buff-Colored Melting Point: 300° Beilstein Reference: 3647189 Merck Reference: 14,2241 MDL No.: MFCD00150612

Formula: (HO)₂C₁₀H₄(SO₃Na)₂·2H₂O
MW: 400.29 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00150612
CAS Number: 5808-22-0
EINECS: 204-972-9
Merck Index: 14,2241

Order Now

Specification Test Results

Solution Pass Test
Appearance Off-white to buff-colored
Sensitivity to nitrate Pass Test
Sensitivity to formaldehyde Pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR