Calcium iodide, anhydrous ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
22395-14
22395-22
CAAA22395-14EA
164.32
CAD
CAAA22395-14
CAAA22395-22
Calcium iodide, anhydrous ≥99.5% (metals basis)
Calcium iodide
MDL: MFCD00010910
Freely soluble in water, methanol, ethanol, acetone. Insoluble in ether
Purity: 99.5%
CAS No.: 10102-68-8
Molecular Formula: CaI2
Formula Weight: 293.89
Physical Form: -20 Mesh Powder
Density: 3.956
Appearance: White to pale yellow
Odor: Odorless
Melting Point: 784°
Boiling Point: 1100°
Solubility: Freely soluble in water, methanol, ethanol, acetone. Insoluble in ether
Merck Reference: 14,1677
MDL No.: MFCD00010910
Formula:
CaI₂ Boiling Pt: 1100 °C Melting Pt: 784 °C Density: 3.956 Storage Temperature: Ambient |
MDL Number:
MFCD00010910 CAS Number: 10102-68-8 EINECS: 233-276-8 Merck Index: 14,1677 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...