Sodium tetrachloroaluminate
Supplier: Thermo Scientific Chemicals
|
Danger
|
22375-14
22375-22
CAAA22375-14EA
489.8
CAD
CAAA22375-14
CAAA22375-22
Sodium tetrachloroaluminate
Sodium tetrachloroaluminate
MDL: MFCD00050361
Soluble in water
CAS No.: 7784-16-9
Molecular Formula: AlCl4Na
Formula Weight: 191.78
Physical Form: -8 Mesh Powder
Density: 2.010
Sensitivity: Moisture Sensitive
Appearance: Pale yellow
Melting Point: 185°
Solubility: Soluble in water
Merck Reference: 14,8687
UN No.: 3260
MDL No.: MFCD00050361
Formula:
AlCl₄Na Melting Pt: 185 °C Density: 2.010 Storage Temperature: Ambient |
MDL Number:
MFCD00050361 CAS Number: 7784-16-9 EINECS: 232-050-6 UN: 3260 ADR: 8,III Merck Index: 14,8687 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...