Trichloroacetic acid (TCA) ≥99% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
TCA
22156-18
22156-30
22156-A1
CAAA22156-18EA
57.58
CAD
CAAA22156-18
CAAA22156-30
CAAA22156-A1
Trichloroacetic acid (TCA) ≥99% ACS
Trichloroacetic acid (TCA)
MDL: MFCD00004177
Beilstein Registry No.: 970119
Common Applications: Protein precipitation reagent.
Fieser: 11194 2425 4520
Purity: 99%
CAS No.: 76-03-9
Molecular Formula: CCl3CO2H
Formula Weight: 163.39
Physical Form: Crystalline
Density: 1.629
pH:
Protein precipitation reagent
Formula:
Cl₃CCOOH Boiling Pt: 196 °C Melting Pt: 54…58 °C Density: 1.629 Flash Pt: 113 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004177 CAS Number: 76-03-9 EINECS: 200-927-2 UN: 1839 ADR: 8,II Merck Index: 14,9627 |
|
Specification Test Results
Clarity of solution | Pass test |
Residue after ignition | 0.03% |
Cl | 0.002% |
NO3 | 0.002% |
PO3 | 5 ppm max. |
SO3 | 0.02% |
Heavy Metals (as Pb) | 0.002% |
Fe | 0.001% |
Substances darkened by sulfuric acid | Pass test |
Insoluble matter | 0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...