Lithium chloride monohydrate ≥99.95% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lithium chloride hydrate, Hydrochloric acid lithium salt monohydrate

22151-14 22151-22
CAAA22151-14EA 192.76 CAD
CAAA22151-14 CAAA22151-22
Lithium chloride monohydrate ≥99.95% (metals basis)
Lithium chloride monohydrate

MDL: MFCD00011078


Purity: 99.95% CAS No.: 16712-20-2 Molecular Formula: LiCl.H2O Formula Weight: 60.41 (42.39anhy) Physical Form: Crystalline Density: 1.78 Melting Point: 98° -H2O Merck Reference: 14,5528 MDL No.: MFCD00011078

Formula: LiCl
MW: 60.41 g/mol
Melting Pt: >98 °C (-H₂O)
Density: 1.78
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 14,5528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR