Calcium peroxide ∼65%

Supplier: Thermo Scientific Chemicals
Danger

21157-22 21157-36
CAAA21157-22EA 114.39 CAD
CAAA21157-22 CAAA21157-36
Calcium peroxide ∼65%
Calcium peroxide

Rubber stabilizer


CAS No.: 1305-79-9 Molecular Formula: CaO2 Formula Weight: 72.08 Physical Form: Powder Sensitivity: Moisture Sensitive Odor: Odorless Merck Reference: 14,1689 UN No.: 1457 MDL No.: MFCD00015988


Rubber stabilizer

Formula: CaO₂
Melting Pt: 692 °C
Density: 2.919…2.921 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015988
CAS Number: 1305-79-9
EINECS: 215-139-4
UN: 1457
ADR: 5.1,II
Merck Index: 14,1689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR