Cerium trifluoride ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

21118-14 21118-22 21118-36
CAAA21118-14EA 123.08 CAD
CAAA21118-14 CAAA21118-22 CAAA21118-36
Cerium trifluoride ≥99.9% (REO, rare earth oxide basis)
Cerium trifluoride

MDL: MFCD00010932 Practically insoluble in, but slowly hydrolyzed by water


Purity: 99.9% CAS No.: 7758-88-5 Molecular Formula: CeF3 Formula Weight: 197.12 Physical Form: Powder Density: 6.16 Melting Point: 1460° Boiling Point: ca 2300° Solubility: Practically insoluble in, but slowly hydrolyzed by water Merck Reference: 14,1998 MDL No.: MFCD00010932

Formula: CeF₃
Boiling Pt: ∼2300 °C
Melting Pt: 1460 °C
Density: 6.16
Storage Temperature: Ambient
MDL Number: MFCD00010932
CAS Number: 7758-88-5
EINECS: 231-841-3
Merck Index: 14,1998

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR