Cerium trichloride, anhydrous ≥99.5% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
21112-09
21112-18
21112-30
CAAA21112-09EA
135.25
CAD
CAAA21112-09
CAAA21112-18
CAAA21112-30
Cerium trichloride, anhydrous ≥99.5% (REO, rare earth oxide basis)
Cerium trichloride
Incandescent gas mantles, spectrography, polymerization of olefins
Purity: 99.5%
CAS No.: 7790-86-5
Molecular Formula: CeCl3
Formula Weight: 246.48
Physical Form: Powder
Density: 3.92
Appearance: White to pale grey
Odor: Odorless
Melting Point: 848°
Boiling Point: 1727°
Solubility: Soluble in water, alcohol, acetone, acids
Merck Reference: 14,1997
MDL No.: MFCD00010929
Incandescent gas mantles, spectrography, polymerization of olefins
Formula:
CeCl₃ Boiling Pt: 1727 °C Melting Pt: 848 °C Density: 3.92 Storage Temperature: Ambient |
MDL Number:
MFCD00010929 CAS Number: 7790-86-5 EINECS: 232-227-8 Merck Index: 14,1997 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...