Tetramethylammonium hydroxide 25% (w/w) in aqueous solution , 99,9999% (metals basis), Electronic Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
20932-AK
20932-K2
20932-LU
20932-LV
CAAA20932-AKEA
92.11
CAD
CAAA20932-AK
CAAA20932-K2
CAAA20932-LU
CAAA20932-LV
Tetramethylammonium hydroxide 25% (w/w) in aqueous solution , 99,9999% (metals basis), Electronic Grade
Tetramethylammonium hydroxide
MDL: MFCD00008280
Beilstein Registry No.: 3558708
Purity: 99.9999%
CAS No.: 75-59-2
Molecular Formula: C4H13NO
Formula Weight: 91.15
Physical Form: Liquid
Density: 1.016
Refractive Index: 1.3840
Sensitivity: Air Sensitive
Appearance: Colorless to pale yellow
Boiling Point: ca 102°
Beilstein Reference: 3558708
Merck Reference: 14,9224
UN No.: 1835
MDL No.: MFCD00008280
Formula:
C₄H₁₃NO Boiling Pt: ∼102 °C Density: 1.016 Storage Temperature: Ambient |
MDL Number:
MFCD00008280 CAS Number: 75-59-2 EINECS: 200-882-9 UN: 1835 ADR: 8,II Merck Index: 14,9224 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...