Sodium trifluoromethanesulfonate
Supplier: Thermo Scientific Chemicals
|
Warning
|
20837-04
20837-09
20837-18
CAAA20837-04EA
45.5
CAD
CAAA20837-04
CAAA20837-09
CAAA20837-18
Sodium trifluoromethanesulfonate
Sodium trifluoromethanesulphonate
MDL: MFCD00061607
Beilstein Registry No.: 3728797
CAS No.: 2926-30-9
Molecular Formula: CF3NaO3S
Formula Weight: 172.06
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 253-255°
Beilstein Reference: 3728797
MDL No.: MFCD00061607
Formula:
CF₃NaO₃S Melting Pt: 253…255 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061607 CAS Number: 2926-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...