Copper(II)-2-ethylhexanoate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Ethylhexanoic acid copper (II) salt, Copper(II) 2-ethylhexanoate, 2-Ethylcaproic acid copper (II) salt, Copper bis(2-ethylhexanoate)

20539-09 20539-18
CAAA20539-09EA 110.76 CAD
CAAA20539-09 CAAA20539-18
Copper(II)-2-ethylhexanoate ≥98%
Copper (II) 2-ethylhexanoate

MDL: MFCD00015695


CAS No.: 149-11-1 Molecular Formula: C16H30CuO4 Formula Weight: 349.96 Physical Form: Powder/Lump Appearance: Blue-green Melting Point: 252° dec. MDL No.: MFCD00015695

Formula: C₁₆H₃₀CuO₄
MW: 349.96
Melting Pt: 252 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00015695
CAS Number: 149-11-1
EINECS: 205-731-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR