1-Butylpyridinium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
19740-04
19740-09
19740-18
CAAA19740-04EA
74.73
CAD
CAAA19740-04
CAAA19740-09
CAAA19740-18
1-Butylpyridinium chloride 98%
1-Butylpyridinium chloride
MDL: MFCD00060047
Purity: 98%
CAS No.: 1124-64-7
Molecular Formula: C9H14ClN
Formula Weight: 171.67
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00060047
Formula:
C₉H₁₄ClN Melting Pt: 131…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060047 CAS Number: 1124-64-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...