Trichloroethylene ≥99.5% stabilized ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00000838
Beilstein Registry No.: 1736782
Miscible with ether, alcohol, chloroform
Fieser: 9,479 10,414 11,552 19,359
Purity: 99.5% min
CAS No.: 79-01-6
Molecular Formula: ClCH=CCl2
Formula Weight: 131.39
Flash Point: None
Physical Form: Liquid
Density: 1.46
Refractive Index: 1.476
Odor: Chloroform-like
Melting Point: -85°
Boiling Point: 87°
Solubility: Miscible with ether, alcohol, chloroform
Beilstein Reference: 1736782
Merck Reference: 14,9639
UN No.: 1710
MDL No.: MFCD00000838
Formula:
ClCH=CCl₂ Boiling Pt: 87 °C Melting Pt: –85 °C Density: 1.46 Storage Temperature: Ambient |
MDL Number:
MFCD00000838 CAS Number: 79-01-6 EINECS: 201-167-4 UN: 1710 ADR: 6.1,III Merck Index: 14,9639 |
|
Specification Test Results
Color (APHA) | 10 |
Evaporation residue | 0.001% |
Titratable acid | 0.0001meq/g |
Titratable base | 0.0003meq/g |
H2O | 0.02% |
Heavy Metals (as Pb) | 1 ppm |
Free halogens | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...