1,2,4-Trichlorobenzene ≥99% for spectrophotometry

Supplier: Thermo Scientific Chemicals
Warning

19390-AK 19390-K2 19390-K7
CAAA19390-AKEA 63.2 CAD
CAAA19390-AK CAAA19390-K2 CAAA19390-K7
1,2,4-Trichlorobenzene ≥99% for spectrophotometry
1,2,4-Trichlorobenzene

MDL: MFCD00000547


Purity: 99% min CAS No.: 120-82-1 Molecular Formula: C6H3Cl3 Formula Weight: 181.45 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.454 Refractive Index: 1.5720 Melting Point: 17° Boiling Point: 213° Merck Reference: 14,9631 UN No.: 2321 MDL No.: MFCD00000547

Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 214.4
Melting Pt: 17
Density: 1.454
Flash Pt: 110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000547
CAS Number: 120-82-1
EINECS: 204-428-0
UN: 2321
ADR: 6.1 III
Merck Index: 14,9631

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR