Pyridine ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
19378-AP
19378-K2
19378-K7
19378-M1
CAAA19378-APEA
146.62
CAD
CAAA19378-AP
CAAA19378-K2
CAAA19378-K7
CAAA19378-M1
Pyridine ≥99.0% ACS
Pyridine
MDL: MFCD00011732
Beilstein Registry No.: 103233
Fieser: 1,958 2,349 4,414 6,497 11,448 12,416
Purity: 99.0% min
CAS No.: 110-86-1
Molecular Formula: C5H5N
Formula Weight: 79.10
Flash Point: 20°(68°F)
Physical Form: Liquid
Density: 0.978
Refractive Index: 1.5100
Appearance: Colorless to pale yellow
Odor: Unpleasant
Melting Point: -42°
Boiling Point: 115°
Beilstein Reference: 103233
Merck Reference: 14,7970
UN No.: 1282
MDL No.: MFCD00011732
Formula:
C₅H₅N Boiling Pt: 115 °C Melting Pt: –42 °C Density: 0.978 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3,II Merck Index: 14,7970 |
|
Specification Test Results
Evaporation residue | 0.002% |
H2O | 0.1% |
Cl | 0.001% |
SO | 0.00001 |
NH3 | 0.002% |
Cu | 5 ppm |
Reducing substances | Pass test |
Solubility in water | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...