Tetrabutylammonium chloride xhydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
19347-06
19347-14
19347-22
CAAA19347-06EA
58.62
CAD
CAAA19347-06
CAAA19347-14
CAAA19347-22
Tetrabutylammonium chloride xhydrate
Tetrabutylammonium chloride hydrate
MDL: MFCD00011635
CAS No.: 37451-68-6
Molecular Formula: [CH3(CH2)3]4NCl.xH2O
Formula Weight: 277.92(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: 73-75°
MDL No.: MFCD00011635
Formula:
C₁₆H₃₆ClN·xH₂O Melting Pt: 73…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011635 CAS Number: 37451-68-6 EINECS: 214-195-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...