Calcium hydride 90-95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
19106-14
19106-22
CAAA19106-14EA
71.42
CAD
CAAA19106-14
CAAA19106-22
CAAA19106-36
Calcium hydride 90-95%
Calcium hydride
MDL: MFCD00010897
Fieser: 1,105 18,83
Purity: 90-95%
CAS No.: 7789-78-8
Molecular Formula: CaH2
Formula Weight: 42.10
Physical Form: 2mm & down
Density: 1.900
Melting Point: 600° dec.
Merck Reference: 14,1672
UN No.: 1404
MDL No.: MFCD00010897
Formula:
H₂Ca Melting Pt: 600 °C (dec.) Density: 1.900 Storage Temperature: Ambient |
MDL Number:
MFCD00010897 CAS Number: 7789-78-8 EINECS: 232-189-2 UN: 1404 ADR: 4.3,I Merck Index: 14,1672 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...