Selenic acid 40% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

18851-22 18851-36
CAAA18851-22EA 353.92 CAD
CAAA18851-22 CAAA18851-36
Selenic acid 40% in aqueous solution
Selenic acid

MDL: MFCD00012191


CAS No.: 7783-08-6 Molecular Formula: H2O4Se Formula Weight: 144.97(anhy) Physical Form: Liquid Density: 1.41 Appearance: Colorless Merck Reference: 14,8429 UN No.: 2922 MDL No.: MFCD00012191

Formula: H₂O₄Se
MW: 144.97
Boiling Pt: 260
Melting Pt: 58
Density: 1.41
Storage Temperature: Ambient
MDL Number: MFCD00012191
CAS Number: 7783-08-6
EINECS: 231-979-4
UN: 2922
ADR: 8 II
Merck Index: 14,8429

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR