Nickel(II) acetylacetonate, anhydrous ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
18811-06
18811-14
18811-22
18811-36
CAAA18811-06EA
54.04
CAD
CAAA18811-06
CAAA18811-14
CAAA18811-22
CAAA18811-36
Nickel(II) acetylacetonate, anhydrous ≥95%
Nickel (II) acetylacetonate
Catalyst and U.V. stabilizer
Purity: 95%
CAS No.: 3264-82-2
Molecular Formula: C10H14NiO4
Formula Weight: 256.91
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Green
Odor: Odorless
Melting Point: ca 230° dec.
Solubility: Soluble in water, alcohol, toluene
Beilstein Reference: 4157970
Merck Reference: 14,6500
MDL No.: MFCD00000024
Catalyst and U.V. stabilizer
Formula:
C₁₀H₁₄NiO₄ Boiling Pt: 220 °C (1013 hPa) Melting Pt: ∼ 230 °C (dec.) Density: 1.455 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000024 CAS Number: 3264-82-2 EINECS: 221-875-7 Merck Index: 14,6500 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...