Tin selenide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

18781-06 18781-14
CAAA18781-06EA 199.08 CAD
CAAA18781-06 CAAA18781-14
Tin selenide ≥99.999% (metals basis)
Tin selenide

MDL: MFCD00054049 Insoluble in water. Soluble in aqua regia, alkali sulfides, and alkali selenides


Purity: 99.999% CAS No.: 1315-06-6 Molecular Formula: SnSe Formula Weight: 197.65 Physical Form: 3-12mm Pieces Melting Point: 861° UN No.: 3077 MDL No.: MFCD00054049

Formula: SeSn
Melting Pt: 861 °C
Density: 6.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054049
CAS Number: 1315-06-6
EINECS: 215-257-6
UN: 3077
ADR: 9,III
Merck Index: 14,8789

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR