Yttrium(III) chloride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

18682-09 18682-18 18682-30
CAAA18682-09EA 140.78 CAD
CAAA18682-09 CAAA18682-18 CAAA18682-30
Yttrium(III) chloride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Yttrium (III) chloride

MDL: MFCD00011470


Purity: 99.9% CAS No.: 10361-92-9 Molecular Formula: Cl3Y Formula Weight: 195.26 Physical Form: -20 Mesh Density: 2.67 Appearance: White to pale yellow Odor: Odorless Melting Point: 721° Merck Reference: 14,10107 MDL No.: MFCD00011470

Formula: Cl₃Y
MW: 195.26 g/mol
Boiling Pt: 1488 °C (1013 hPa)
Melting Pt: 721 °C
Density: 2.67
Storage Temperature: Ambient
MDL Number: MFCD00011470
CAS Number: 10361-92-9
EINECS: 233-801-0
Merck Index: 14,10107

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR