Holmium(III) chloride, anhydrous ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Holmium trichloride

18681-06 18681-14
CAAA18681-06EA 211.72 CAD
CAAA18681-06 CAAA18681-14
Holmium(III) chloride, anhydrous ≥99.9% (metals basis)
Holmium(III) chloride

MDL: MFCD00011051


Purity: 99.9% CAS No.: 10138-62-2 Molecular Formula: HoCl3 Formula Weight: 271.29 Physical Form: -20 Mesh Density: 3.7 Melting Point: 718° Merck Reference: 14,4727 MDL No.: MFCD00011051

Formula: Cl₃Ho
MW: 271.29 g/mol
Boiling Pt: 1500 °C (1013 hPa)
Melting Pt: 718 °C
Density: 3.7
Storage Temperature: Ambient
MDL Number: MFCD00011051
CAS Number: 10138-62-2
EINECS: 233-387-1
Merck Index: 14,4727

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR