3-Methoxypropene 94%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Allyl methyl ether

18652-09 18652-18
CAAA18652-09EA 218.04 CAD
CAAA18652-09 CAAA18652-18
3-Methoxypropene 94%
3-Methoxypropene

MDL: MFCD00008649


Purity: 94% CAS No.: 627-40-7 Molecular Formula: CH2=CHCH2OCH3 Formula Weight: 72.10 Flash Point: -23°(-94°F) Physical Form: Liquid Density: 0.77 Refractive Index: 1.3803 Odor: Ether-like Boiling Point: 42-43° UN No.: 3271 MDL No.: MFCD00008649

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 42…43 °C
Density: 0,77
Flash Pt: –23 °C (–94 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008649
CAS Number: 627-40-7
EINECS: 210-997-6
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR