Silver methanesulfonate
Supplier: Thermo Scientific Chemicals
|
Danger
|
18641-09
18641-18
CAAA18641-09EA
199.08
CAD
CAAA18641-09
CAAA18641-18
Silver methanesulfonate
Silver methanesulphonate
MDL: MFCD00064795
CAS No.: 2386-52-9
Molecular Formula: CH3AgO3S
Formula Weight: 202.97
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White to grey
Odor: Odorless
UN No.: 3077
MDL No.: MFCD00064795
Formula:
CH₃AgO₃S Melting Pt: 263 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064795 CAS Number: 2386-52-9 EINECS: 219-199-2 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...